Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM29
Entrez Gene ID
23650
Gene Name
tripartite motif containing 29
Gene Aliases
ATDC
Chromosome Location
Chr.11: 120111286 - 120138179 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012101.3 NP_036233.2 8 - 9 1831 75
XM_005271490.1 XP_005271547.1 8 - 9 1008 75
XM_011542729.1 XP_011541031.1 9 - 10 1957 75
XM_011542732.1 XP_011541034.1 9 - 10 1954 75
XM_011542733.1 XP_011541035.1 9 - 10 1110 75
XM_017017453.1 XP_016872942.1 9 - 10 1266 75
XM_017017454.1 XP_016872943.1 8 - 9 1164 75
GenBank mRNA AF230388.1 - 8 - 9 1831 75
AK093404.1 - 4 - 5 901 75
AK122720.1 - 9 - 10 1647 75
AK223090.1 - 8 - 9 1789 75
AK299459.1 - 8 - 9 1109 75
AK299472.1 - 8 - 9 1188 75
AK303969.1 - 8 - 9 1069 75
AL832678.1 - 1 - 2 1773 75
BC010644.1 - 8 - 9 1777 75
BC017352.2 - 8 - 9 1770 75
L24203.1 - 8 - 9 1831 75

View Full Product Details