The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004621.5 | NP_004612.2 | 7 - 8 | 2427 | 113 |
XM_011542968.2 | XP_011541270.1 | 7 - 8 | 2034 | 113 | |
XM_017018221.1 | XP_016873710.1 | 5 - 6 | 2169 | 113 | |
XM_017018222.1 | XP_016873711.1 | 7 - 8 | 1929 | 113 | |
GenBank mRNA | AF080394.2 | - | 7 - 8 | 2019 | 113 |
AJ006276.1 | - | 7 - 8 | 2429 | 113 | |
AJ271066.1 | - | 7 - 8 | 2002 | 113 | |
AJ271067.1 | - | 5 - 6 | 1654 | 113 | |
AJ271068.1 | - | 6 - 7 | 1837 | 113 | |
AK027769.1 | - | 6 - 7 | 1072 | 113 | |
BC093658.1 | - | 7 - 8 | 2066 | 113 | |
BC093660.1 | - | 7 - 8 | 2066 | 113 |