The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004621.5 | NP_004612.2 | 9 - 10 | 2827 | 146 |
XM_011542968.2 | XP_011541270.1 | 9 - 10 | 2434 | 146 | |
XM_017018221.1 | XP_016873710.1 | 7 - 8 | 2569 | 146 | |
XM_017018222.1 | XP_016873711.1 | 9 - 10 | 2329 | 146 | |
GenBank mRNA | AF080394.2 | - | 9 - 10 | 2419 | 146 |
AJ006276.1 | - | 9 - 10 | 2829 | 146 | |
AJ271066.1 | - | 9 - 10 | 2402 | 146 | |
AJ271067.1 | - | 7 - 8 | 2054 | 146 | |
AJ271068.1 | - | 8 - 9 | 2237 | 146 | |
AK027769.1 | - | 8 - 9 | 1472 | 146 | |
BC093658.1 | - | 9 - 10 | 2466 | 146 | |
BC093660.1 | - | 9 - 10 | 2466 | 146 |