Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRPC6
Entrez Gene ID
7225
Gene Name
transient receptor potential cation channel subfamily C member 6
Gene Aliases
FSGS2, TRP6
Chromosome Location
Chr.11: 101451470 - 101584018 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004621.5 NP_004612.2 2 - 3 1366 122
XM_011542968.2 XP_011541270.1 2 - 3 973 122
XM_017018222.1 XP_016873711.1 2 - 3 868 122
GenBank mRNA AF080394.2 - 2 - 3 958 122
AJ006276.1 - 2 - 3 1368 122
AJ271066.1 - 2 - 3 941 122
AJ271068.1 - 2 - 3 941 122
BC093658.1 - 2 - 3 1005 122
BC093660.1 - 2 - 3 1005 122

View Full Product Details