The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168618.1 | NP_001162089.1 | 9 - 10 | 1094 | 67 |
NM_020153.3 | NP_064538.3 | 10 - 11 | 1247 | 67 | |
XM_011542905.2 | XP_011541207.1 | 10 - 11 | 3989 | 67 | |
XM_011542906.2 | XP_011541208.1 | 9 - 10 | 3831 | 67 | |
XM_017018017.1 | XP_016873506.1 | 9 - 10 | 986 | 67 | |
XM_017018018.1 | XP_016873507.1 | 8 - 9 | 957 | 67 | |
GenBank mRNA | AJ249980.1 | - | 9 - 10 | 1021 | 67 |
AK025480.1 | - | 10 - 11 | 1206 | 67 | |
AK289521.1 | - | 9 - 10 | 945 | 67 | |
AL136934.1 | - | 9 - 10 | 929 | 67 | |
BC011647.2 | - | 9 - 10 | 932 | 67 | |
BC012802.2 | - | 9 - 10 | 1025 | 67 | |
BC022856.1 | - | 9 - 10 | 1055 | 67 | |
CR457344.1 | - | 7 - 8 | 675 | 67 |