The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303012.1 | NP_001289941.1 | 7 - 8 | 1201 | 106 |
NM_001303013.1 | NP_001289942.1 | 7 - 8 | 2387 | 106 | |
NM_014638.3 | NP_055453.2 | 7 - 8 | 1395 | 106 | |
XM_011542455.2 | XP_011540757.1 | 8 - 9 | 1437 | 106 | |
XM_011542456.2 | XP_011540758.1 | 8 - 9 | 1394 | 106 | |
XM_017002870.1 | XP_016858359.1 | 8 - 9 | 1426 | 106 | |
XM_017002871.1 | XP_016858360.1 | 8 - 9 | 1397 | 106 | |
XM_017002872.1 | XP_016858361.1 | 8 - 9 | 1426 | 106 | |
XM_017002873.1 | XP_016858362.1 | 8 - 9 | 1426 | 106 | |
XM_017002874.1 | XP_016858363.1 | 8 - 9 | 1426 | 106 | |
GenBank mRNA | AB007919.2 | - | 7 - 8 | 1201 | 106 |
AK122589.1 | - | 5 - 6 | 1494 | 106 | |
BC043358.1 | - | 5 - 6 | 697 | 106 | |
BC128207.1 | - | 5 - 6 | 777 | 106 | |
BC144620.1 | - | 7 - 8 | 2379 | 106 | |
DQ176850.1 | - | 7 - 8 | 1395 | 106 |