The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303012.1 | NP_001289941.1 | 11 - 12 | 1740 | 60 |
NM_001303013.1 | NP_001289942.1 | 11 - 12 | 2926 | 60 | |
NM_014638.3 | NP_055453.2 | 11 - 12 | 1934 | 60 | |
XM_011542455.2 | XP_011540757.1 | 12 - 13 | 1976 | 60 | |
XM_011542456.2 | XP_011540758.1 | 12 - 13 | 1933 | 60 | |
XM_017002870.1 | XP_016858359.1 | 12 - 13 | 1965 | 60 | |
XM_017002871.1 | XP_016858360.1 | 12 - 13 | 1936 | 60 | |
XM_017002872.1 | XP_016858361.1 | 12 - 13 | 1965 | 60 | |
XM_017002873.1 | XP_016858362.1 | 12 - 13 | 1965 | 60 | |
XM_017002874.1 | XP_016858363.1 | 12 - 13 | 1965 | 60 | |
GenBank mRNA | AB007919.2 | - | 11 - 12 | 1740 | 60 |
AK122589.1 | - | 9 - 10 | 2033 | 60 | |
BC043358.1 | - | 9 - 10 | 1236 | 60 | |
BC128207.1 | - | 9 - 10 | 1316 | 60 | |
BC144620.1 | - | 11 - 12 | 2918 | 60 | |
DQ176850.1 | - | 11 - 12 | 1934 | 60 |