Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MDS2
- Entrez Gene ID
-
259283
- Gene Name
- myelodysplastic syndrome 2 translocation associated
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 23627334 - 23640566 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027042.1 |
|
5 - 6 |
732 |
74 |
GenBank mRNA |
AJ310434.2 |
- |
5 - 6 |
732 |
74 |
BC069164.1 |
- |
5 - 6 |
732 |
74 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
inhibitor of DNA binding 3, HLH protein |
ID3 |
HEIR-1, bHLHb25 |
-79.3kb
|
Anti-sense |
Chr. - 23557930 - 23559794 on Build GRCh38 |
Downstream |
ribosomal protein L11 |
RPL11 |
DBA7, GIG34, L11 |
52.0kb
|
Sense |
Chr. - 23691779 - 23696425 on Build GRCh38 |
View Full Product Details