Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RIMS2
Entrez Gene ID
9699
Gene Name
regulating synaptic membrane exocytosis 2
Gene Aliases
OBOE, RAB3IP3, RIM2
Chromosome Location
Chr.8: 103500026 - 104254430 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014677.4 NP_055492.3 14 - 15 2734 90
XM_005251106.3 XP_005251163.1 16 - 17 3270 90
XM_011517398.2 XP_011515700.1 21 - 22 3371 90
XM_017014007.1 XP_016869496.1 21 - 22 3386 90
XM_017014018.1 XP_016869507.1 16 - 17 3129 90
XM_017014022.1 XP_016869511.1 14 - 15 2794 90
XM_017014023.1 XP_016869512.1 20 - 21 3323 90
XM_017014027.1 XP_016869516.1 20 - 21 3182 90
XM_017014029.1 XP_016869518.1 17 - 18 4004 90
XM_017014030.1 XP_016869519.1 16 - 17 3270 90
XM_017014034.1 XP_016869523.1 18 - 19 3911 90
XM_017014035.1 XP_016869524.1 17 - 18 3862 90
XM_017014036.1 XP_016869525.1 14 - 15 2653 90
XM_017014043.1 XP_016869532.1 14 - 15 2794 90
GenBank mRNA AK308831.1 - 14 - 15 2534 90
BC043144.2 - 14 - 15 2650 90
CR749374.1 - 15 - 16 2614 90

View Full Product Details