The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000098.2 | NP_000089.1 | 2 - 3 | 747 | 91 |
XM_005270484.1 | XP_005270541.1 | 2 - 3 | 415 | 91 | |
GenBank mRNA | AK312687.1 | - | 2 - 3 | 351 | 91 |
BC002445.2 | - | 2 - 3 | 356 | 91 | |
BC005172.2 | - | 2 - 3 | 356 | 91 | |
M58581.1 | - | 2 - 3 | 333 | 91 | |
U09648.1 | - | 2 - 3 | 748 | 91 |