The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002439.4 | NP_002430.3 | 15 - 16 | 2330 | 147 |
GenBank mRNA | AK289856.1 | - | 16 - 17 | 2299 | 147 |
BC004177.2 | - | 16 - 17 | 2271 | 147 | |
BC011817.2 | - | 16 - 17 | 2271 | 147 | |
BC017273.2 | - | 15 - 16 | 2253 | 147 | |
BC098436.1 | - | 16 - 17 | 2271 | 147 | |
BC130434.1 | - | 15 - 16 | 2330 | 147 | |
BC130436.1 | - | 15 - 16 | 2330 | 147 | |
J04810.1 | - | 15 - 16 | 2311 | 147 | |
U61981.1 | - | 16 - 17 | 2239 | 147 |