Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MSH3
Entrez Gene ID
4437
Gene Name
mutS homolog 3
Gene Aliases
DUP, MRP1
Chromosome Location
Chr.5: 80654648 - 80876815 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002439.4 NP_002430.3 23 - 24 3383 105
GenBank mRNA AK289856.1 - 24 - 25 3352 105
BC130434.1 - 23 - 24 3383 105
BC130436.1 - 23 - 24 3383 105
J04810.1 - 23 - 24 3364 105
U61981.1 - 24 - 25 3292 105

View Full Product Details