The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005926.2 | NP_005917.2 | 4 - 5 | 802 | 66 |
GenBank mRNA | AK312867.1 | - | 4 - 5 | 778 | 66 |
BC023557.2 | - | 4 - 5 | 737 | 66 | |
BC050742.1 | - | 4 - 5 | 748 | 66 | |
U04209.1 | - | 4 - 5 | 736 | 66 |