Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYAR
Entrez Gene ID
55646
Gene Name
Ly1 antibody reactive
Gene Aliases
ZC2HC2, ZLYAR
Chromosome Location
Chr.4: 4267701 - 4290169 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017816.2 NP_060286.1 1 - 2 134 87
GenBank mRNA AI418609.1 - 1 - 2 134 87
AK000432.1 - 1 - 2 81 87
AK290750.1 - 1 - 2 111 87
BC015796.2 - 1 - 2 96 87

View Full Product Details