Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSPAN2
Entrez Gene ID
10100
Gene Name
tetraspanin 2
Gene Aliases
NET3, TSN2, TSPAN-2
Chromosome Location
Chr.1: 115048011 - 115089501 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308315.1 NP_001295244.1 6 - 7 593 101
NM_005725.5 NP_005716.2 7 - 8 668 101
GenBank mRNA AF054839.1 - 7 - 8 603 101
AK315385.1 - 7 - 8 628 101
BC021675.1 - 7 - 8 633 101
BG697236.1 - 3 - 4 250 101
BQ067895.1 - 7 - 8 680 101
CB993308.1 - 6 - 7 619 101

View Full Product Details