The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177562.1 | NP_001171033.1 | 12 - 13 | 1647 | 90 |
NM_001177563.1 | NP_001171034.1 | 11 - 12 | 1401 | 90 | |
NM_002716.4 | NP_002707.3 | 13 - 14 | 1782 | 90 | |
NM_181699.2 | NP_859050.1 | 13 - 14 | 1782 | 90 | |
NM_181700.1 | NP_859051.1 | 11 - 12 | 1590 | 90 | |
XM_017017958.1 | XP_016873447.1 | 13 - 14 | 1726 | 90 | |
XM_017017959.1 | XP_016873448.1 | 13 - 14 | 1726 | 90 | |
XM_017017960.1 | XP_016873449.1 | 12 - 13 | 1591 | 90 | |
XM_017017961.1 | XP_016873450.1 | 13 - 14 | 1726 | 90 | |
GenBank mRNA | AF087438.1 | - | 13 - 14 | 1727 | 90 |
AF163473.1 | - | 13 - 14 | 1764 | 90 | |
AK292275.1 | - | 13 - 14 | 1702 | 90 | |
AK293388.1 | - | 13 - 14 | 1646 | 90 | |
AK294716.1 | - | 12 - 13 | 1570 | 90 | |
AK296455.1 | - | 11 - 12 | 1339 | 90 | |
AK301705.1 | - | 11 - 12 | 1531 | 90 | |
BC027596.2 | - | 13 - 14 | 1719 | 90 | |
DB527885.1 | - | 1 - 2 | 90 | ||
M65254.1 | - | 13 - 14 | 1620 | 90 |