The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252127.1 | NP_001239056.1 | 15 - 16 | 2095 | 75 |
NM_007347.4 | NP_031373.2 | 15 - 16 | 2072 | 75 | |
XM_005254264.3 | XP_005254321.1 | 15 - 16 | 1890 | 75 | |
XM_006720447.3 | XP_006720510.1 | 15 - 16 | 2250 | 75 | |
XM_017022042.1 | XP_016877531.1 | 15 - 16 | 2105 | 75 | |
GenBank mRNA | AB030653.1 | - | 15 - 16 | 1995 | 75 |
AF155156.2 | - | 15 - 16 | 1998 | 75 | |
BC126308.1 | - | 15 - 16 | 2071 | 75 | |
BC130466.1 | - | 15 - 16 | 2071 | 75 | |
CR749604.1 | - | 15 - 16 | 1972 | 75 |