The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000338.2 | NP_000329.2 | 14 - 15 | 1997 | 98 |
NM_001184832.1 | NP_001171761.1 | 14 - 15 | 1997 | 98 | |
XM_005254606.1 | XP_005254663.1 | 14 - 15 | 1997 | 98 | |
GenBank mRNA | AK295375.1 | - | 13 - 14 | 1450 | 98 |
AK298275.1 | - | 9 - 10 | 1216 | 98 | |
AK298312.1 | - | 14 - 15 | 1996 | 98 | |
BX647067.1 | - | 9 - 10 | 5920 | 98 | |
BX647484.1 | - | 10 - 11 | 5904 | 98 | |
EF559316.2 | - | 13 - 14 | 1795 | 98 | |
U58130.1 | - | 13 - 14 | 1800 | 98 |