Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC12A1
Entrez Gene ID
6557
Gene Name
solute carrier family 12 member 1
Gene Aliases
BSC1, NKCC2
Chromosome Location
Chr.15: 48206301 - 48304078 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000338.2 NP_000329.2 2 - 3 637 66
NM_001184832.1 NP_001171761.1 2 - 3 637 66
XM_005254606.1 XP_005254663.1 2 - 3 637 66
GenBank mRNA AK298312.1 - 2 - 3 636 66
BC040138.2 - 1 - 2 607 66
EF559316.2 - 1 - 2 435 66
U58130.1 - 1 - 2 440 66

View Full Product Details