The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000338.2 | NP_000329.2 | 9 - 10 | 1432 | 78 |
NM_001184832.1 | NP_001171761.1 | 9 - 10 | 1432 | 78 | |
XM_005254606.1 | XP_005254663.1 | 9 - 10 | 1432 | 78 | |
GenBank mRNA | AK295375.1 | - | 8 - 9 | 885 | 78 |
AK298275.1 | - | 4 - 5 | 651 | 78 | |
AK298312.1 | - | 9 - 10 | 1431 | 78 | |
BX647067.1 | - | 4 - 5 | 5355 | 78 | |
BX647484.1 | - | 5 - 6 | 5339 | 78 | |
EF559316.2 | - | 8 - 9 | 1230 | 78 | |
U58130.1 | - | 8 - 9 | 1235 | 78 |