The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005609.1 | NP_001005609.1 | 2 - 3 | 746 | 79 |
NM_001005612.2 | NP_001005612.2 | 2 - 3 | 746 | 79 | |
NM_001399.4 | NP_001390.1 | 2 - 3 | 746 | 79 | |
XM_006724630.2 | XP_006724693.1 | 2 - 3 | 727 | 79 | |
XM_011530885.2 | XP_011529187.1 | 2 - 3 | 727 | 79 | |
XM_017029336.1 | XP_016884825.1 | 2 - 3 | 727 | 79 | |
GenBank mRNA | AF040628.1 | - | 2 - 3 | 746 | 79 |
AF060999.1 | - | 2 - 3 | 745 | 79 | |
AF061189.1 | - | 2 - 3 | 746 | 79 | |
BC126143.1 | - | 2 - 3 | 696 | 79 | |
BC144049.1 | - | 2 - 3 | 696 | 79 | |
BC144051.1 | - | 2 - 3 | 696 | 79 |