Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRMD1
Entrez Gene ID
79981
Gene Name
FERM domain containing 1
Gene Aliases
bA164L23.1
Chromosome Location
Chr.6: 168055745 - 168093007 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_110312.1 7 - 8 1085 73
GenBank mRNA AK125963.1 - 7 - 8 1057 73
AL133077.1 - 1 - 2 84 73

View Full Product Details