Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ATP6V0A1
- Entrez Gene ID
-
535
- Gene Name
- ATPase H+ transporting V0 subunit a1
- Gene Aliases
-
ATP6N1, ATP6N1A, Stv1, VPP1, Vph1, a1
- Chromosome Location
-
Chr.17: 42458844 - 42522579 on Build GRCh38
View Full Product Details