Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEMA3C
Entrez Gene ID
10512
Gene Name
semaphorin 3C
Gene Aliases
SEMAE, SemE
Chromosome Location
Chr.7: 80742538 - 80919351 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006379.3 NP_006370.1 2 - 3 663 80
GenBank mRNA AB000220.1 - 2 - 3 663 80
AK304294.1 - 2 - 3 234 80
AK312478.1 - 2 - 3 374 80
BC030690.1 - 2 - 3 466 80

View Full Product Details