Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC17A1
Entrez Gene ID
6568
Gene Name
solute carrier family 17 member 1
Gene Aliases
NAPI-1, NPT-1, NPT1
Chromosome Location
Chr.6: 25723744 - 25832108 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005074.3 NP_005065.2 9 - 10 1147 62
XM_011514818.2 XP_011513120.2 9 - 10 1184 62
XM_011514819.2 XP_011513121.2 8 - 9 1097 62
XM_011514820.2 XP_011513122.2 8 - 9 1022 62
XM_011514821.2 XP_011513123.1 7 - 8 901 62
XM_017011199.1 XP_016866688.1 9 - 10 1184 62
XM_017011200.1 XP_016866689.1 9 - 10 1184 62
XM_017011201.1 XP_016866690.1 9 - 10 1184 62
XM_017011202.1 XP_016866691.1 8 - 9 1112 62
GenBank mRNA AK290783.1 - 9 - 10 1129 62
AK308035.1 - 9 - 10 1129 62
AY780791.1 - 7 - 8 869 62
BC101745.1 - 8 - 9 1084 62
BC101747.1 - 8 - 9 1084 62
D28532.1 - 8 - 9 1028 62
X71355.1 - 8 - 9 1043 62

View Full Product Details