The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278.4 | NP_001269.3 | 12 - 13 | 1429 | 133 |
NM_001320928.1 | NP_001307857.1 | 12 - 13 | 1429 | 133 | |
XM_017015611.1 | XP_016871100.1 | 12 - 13 | 1429 | 133 | |
XM_017015612.1 | XP_016871101.1 | 12 - 13 | 1429 | 133 | |
XM_017015613.1 | XP_016871102.1 | 2 - 3 | 244 | 133 | |
GenBank mRNA | AB209028.1 | - | 12 - 13 | 1383 | 133 |
AF009225.1 | - | 12 - 13 | 1386 | 133 | |
AF012890.1 | - | 12 - 13 | 1351 | 133 | |
AF080157.1 | - | 12 - 13 | 1446 | 133 | |
BC092514.1 | - | 12 - 13 | 1367 | 133 | |
BM800404.1 | - | - | 614 | 133 | |
U22512.1 | - | 11 - 12 | 1259 | 133 |