The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000292.2 | NP_000283.1 | 16 - 17 | 2145 | 70 |
XM_005274548.4 | XP_005274605.1 | 16 - 17 | 1899 | 70 | |
XM_005274550.4 | XP_005274607.1 | 16 - 17 | 1899 | 70 | |
XM_006724496.3 | XP_006724559.1 | 16 - 17 | 1899 | 70 | |
XM_006724498.3 | XP_006724561.1 | 15 - 16 | 1569 | 70 | |
XM_011545537.2 | XP_011543839.1 | 15 - 16 | 1800 | 70 | |
XM_011545538.2 | XP_011543840.1 | 9 - 10 | 943 | 70 | |
XM_017029580.1 | XP_016885069.1 | 13 - 14 | 1274 | 70 | |
GenBank mRNA | AK289996.1 | - | 16 - 17 | 1866 | 70 |
BC014036.2 | - | 16 - 17 | 1932 | 70 | |
D38616.1 | - | 16 - 17 | 1886 | 70 | |
X80497.1 | - | 16 - 17 | 1841 | 70 |