The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000292.2 | NP_000283.1 | 26 - 27 | 3338 | 68 |
XM_005274548.4 | XP_005274605.1 | 26 - 27 | 3092 | 68 | |
XM_005274550.4 | XP_005274607.1 | 26 - 27 | 3092 | 68 | |
XM_006724496.3 | XP_006724559.1 | 26 - 27 | 3092 | 68 | |
XM_006724498.3 | XP_006724561.1 | 25 - 26 | 2762 | 68 | |
XM_011545537.2 | XP_011543839.1 | 25 - 26 | 2993 | 68 | |
XM_011545538.2 | XP_011543840.1 | 19 - 20 | 2136 | 68 | |
XM_017029580.1 | XP_016885069.1 | 23 - 24 | 2467 | 68 | |
GenBank mRNA | AK289996.1 | - | 26 - 27 | 3059 | 68 |
AY429559.1 | - | 1 - 2 | 88 | 68 | |
BC014036.2 | - | 26 - 27 | 3125 | 68 | |
D38616.1 | - | 26 - 27 | 3079 | 68 | |
X80497.1 | - | 26 - 27 | 3034 | 68 |