The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000292.2 | NP_000283.1 | 29 - 30 | 3539 | 59 |
GenBank mRNA | AK289996.1 | - | 29 - 30 | 3260 | 59 |
BC014036.2 | - | 29 - 30 | 3326 | 59 | |
D38616.1 | - | 29 - 30 | 3280 | 59 | |
X73875.1 | - | 3 - 4 | 182 | 59 | |
X80497.1 | - | 29 - 30 | 3235 | 59 |