The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286076.1 | NP_001273005.1 | 4 - 5 | 842 | 58 |
NM_001297558.1 | NP_001284487.1 | 4 - 5 | 498 | 58 | |
NM_002946.4 | NP_002937.1 | 4 - 5 | 691 | 58 | |
GenBank mRNA | AK298759.1 | - | 4 - 5 | 451 | 58 |
AK300691.1 | - | 4 - 5 | 584 | 58 | |
AK312956.1 | - | 4 - 5 | 455 | 58 | |
BC001630.1 | - | 4 - 5 | 428 | 58 | |
BC012157.1 | - | 4 - 5 | 480 | 58 | |
BC021257.2 | - | 4 - 5 | 619 | 58 | |
BX333932.2 | - | 4 - 5 | 420 | 58 | |
CR450348.1 | - | 3 - 4 | 337 | 58 | |
J05249.1 | - | 4 - 5 | 414 | 58 |