The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012096.2 | NP_036228.1 | 8 - 9 | 761 | 129 |
XM_011533583.2 | XP_011531885.1 | 9 - 10 | 814 | 129 | |
GenBank mRNA | AB037849.2 | - | 8 - 9 | 614 | 129 |
AF169797.1 | - | 8 - 9 | 672 | 129 | |
AF424738.1 | - | 8 - 9 | 670 | 129 | |
AK299008.1 | - | 9 - 10 | 803 | 129 | |
AK310716.1 | - | 8 - 9 | 709 | 129 | |
BC028599.2 | - | 8 - 9 | 761 | 129 |