The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307944.1 | NP_001294873.1 | 2 - 3 | 630 | 75 |
NM_145052.3 | NP_659489.1 | 2 - 3 | 630 | 75 | |
GenBank mRNA | AK056354.1 | - | 2 - 3 | 599 | 75 |
AK293024.1 | - | 2 - 3 | 526 | 75 | |
AK309951.1 | - | 2 - 3 | 604 | 75 | |
AY643714.1 | - | 2 - 3 | 616 | 75 | |
BC015116.1 | - | 2 - 3 | 565 | 75 |