The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300793.1 | NP_001287722.1 | 11 - 12 | 1679 | 64 |
NM_001300794.1 | NP_001287723.1 | 11 - 12 | 1676 | 64 | |
NM_153265.2 | NP_694997.2 | 11 - 12 | 1676 | 64 | |
XM_005273878.3 | XP_005273935.1 | 11 - 12 | 1934 | 64 | |
XM_006718489.3 | XP_006718552.1 | 11 - 12 | 2051 | 64 | |
XM_006718490.3 | XP_006718553.1 | 11 - 12 | 2051 | 64 | |
XM_006718491.3 | XP_006718554.1 | 11 - 12 | 2048 | 64 | |
XM_011544896.2 | XP_011543198.1 | 11 - 12 | 2048 | 64 | |
XM_017017480.1 | XP_016872969.1 | 11 - 12 | 1422 | 64 | |
XM_017017481.1 | XP_016872970.1 | 10 - 11 | 1308 | 64 | |
XM_017017482.1 | XP_016872971.1 | 11 - 12 | 1422 | 64 | |
XM_017017483.1 | XP_016872972.1 | 11 - 12 | 1479 | 64 | |
GenBank mRNA | AK093146.1 | - | 10 - 11 | 1351 | 64 |
AK128679.1 | - | 10 - 11 | 2093 | 64 | |
AK296901.1 | - | 11 - 12 | 1441 | 64 | |
AK310579.1 | - | 13 - 14 | 1502 | 64 | |
AL833388.1 | - | 8 - 9 | 931 | 64 | |
BC075824.1 | - | 10 - 11 | 1282 | 64 | |
BC108280.1 | - | 11 - 12 | 1460 | 64 |