The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001522.2 | NP_001513.2 | 18 - 19 | 3516 | 112 |
GenBank mRNA | BC144241.1 | - | 19 - 20 | 3463 | 112 |
L37378.1 | - | 18 - 19 | 3539 | 112 |