The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014578.3 | NP_055393.1 | 3 - 4 | 417 | 77 |
GenBank mRNA | AF498973.1 | - | 3 - 4 | 332 | 77 |
BC001338.1 | - | 3 - 4 | 385 | 77 | |
BM761362.1 | - | 3 - 4 | 417 | 77 |