The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242314.1 | NP_001229243.1 | 3 - 4 | 763 | 114 |
NM_004721.4 | NP_004712.1 | 2 - 3 | 878 | 114 | |
XM_011513310.2 | XP_011511612.1 | 2 - 3 | 1036 | 114 | |
XM_017007456.1 | XP_016862945.1 | 3 - 4 | 774 | 114 | |
GenBank mRNA | AB001872.1 | - | 2 - 3 | 744 | 114 |
AK296961.1 | - | 2 - 3 | 806 | 114 | |
AK312714.1 | - | 3 - 4 | 728 | 114 |