The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000228.2 | NP_000219.2 | 18 - 19 | 2848 | 67 |
NM_001017402.1 | NP_001017402.1 | 17 - 18 | 3094 | 67 | |
NM_001127641.1 | NP_001121113.1 | 18 - 19 | 2822 | 67 | |
XM_005273124.4 | XP_005273181.1 | 18 - 19 | 3030 | 67 | |
XM_017001272.1 | XP_016856761.1 | 17 - 18 | 3258 | 67 | |
GenBank mRNA | AY035783.1 | - | 17 - 18 | 2704 | 67 |
BC075838.1 | - | 17 - 18 | 2879 | 67 | |
D37766.1 | - | 18 - 19 | 2785 | 67 | |
KU178159.1 | - | 17 - 18 | 2704 | 67 | |
L25541.1 | - | 18 - 19 | 2818 | 67 |