The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001280.2 | NP_001271.1 | 5 - 6 | 584 | 65 |
NM_001300815.1 | NP_001287744.1 | 5 - 6 | 586 | 65 | |
NM_001300829.1 | NP_001287758.1 | 5 - 6 | 586 | 65 | |
NR_023312.2 | 5 - 6 | 645 | 65 | ||
NR_023313.2 | 4 - 5 | 477 | 65 | ||
XM_006722637.1 | XP_006722700.1 | 5 - 6 | 647 | 65 | |
XM_011527668.1 | XP_011525970.1 | 5 - 6 | 520 | 65 | |
XM_017026237.1 | XP_016881726.1 | 5 - 6 | 647 | 65 | |
GenBank mRNA | AB451225.1 | - | 4 - 5 | 435 | 65 |
AB451348.1 | - | 4 - 5 | 435 | 65 | |
AF021336.1 | - | 5 - 6 | 517 | 65 | |
AK094781.1 | - | 4 - 5 | 410 | 65 | |
AK095781.1 | - | 3 - 4 | 705 | 65 | |
AK128423.1 | - | 4 - 5 | 499 | 65 | |
AK304185.1 | - | 4 - 5 | 663 | 65 | |
AK304470.1 | - | 5 - 6 | 418 | 65 | |
BC000403.2 | - | 5 - 6 | 496 | 65 | |
BC000901.1 | - | 5 - 6 | 501 | 65 | |
BM706055.1 | - | 5 - 6 | 498 | 65 | |
BT007210.1 | - | 4 - 5 | 435 | 65 | |
D78134.1 | - | 5 - 6 | 515 | 65 | |
DA433715.1 | - | 5 - 6 | 521 | 65 |