Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FRZB
Entrez Gene ID
2487
Gene Name
frizzled-related protein
Gene Aliases
FRE, FRITZ, FRP-3, FRZB-1, FRZB-PEN, FRZB1, FZRB, OS1, SFRP3, SRFP3, hFIZ
Chromosome Location
Chr.2: 182833275 - 182866770 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001463.3 NP_001454.2 4 - 5 1009 104
GenBank mRNA AK130009.1 - 5 - 6 661 104
AK312741.1 - 4 - 5 888 104
BC027855.1 - 4 - 5 861 104
BT019883.1 - 4 - 5 791 104
U24163.1 - 4 - 5 999 104
U68057.1 - 4 - 5 851 104
U91903.1 - 4 - 5 860 104

View Full Product Details