Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXJ2
Entrez Gene ID
55810
Gene Name
forkhead box J2
Gene Aliases
FHX
Chromosome Location
Chr.12: 8032680 - 8055522 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018416.2 NP_060886.1 10 - 11 2721 70
XM_011520760.2 XP_011519062.1 11 - 12 2895 70
XM_011520761.2 XP_011519063.1 12 - 13 2996 70
XM_017019640.1 XP_016875129.1 10 - 11 1559 70
GenBank mRNA AF155132.1 - 10 - 11 2125 70
AL161978.1 - 2 - 3 265 70
BC126396.1 - 10 - 11 1691 70
BC136305.1 - 10 - 11 1691 70

View Full Product Details