Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXJ2
Entrez Gene ID
55810
Gene Name
forkhead box J2
Gene Aliases
FHX
Chromosome Location
Chr.12: 8032680 - 8055522 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018416.2 NP_060886.1 2 - 3 1419 107
XM_011520760.2 XP_011519062.1 2 - 3 1502 107
XM_011520761.2 XP_011519063.1 3 - 4 1603 107
GenBank mRNA AF155132.1 - 2 - 3 823 107
AF155133.1 - 2 - 3 823 107
BC126396.1 - 2 - 3 389 107
BC136305.1 - 2 - 3 389 107

View Full Product Details