Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FOXJ2
Entrez Gene ID
55810
Gene Name
forkhead box J2
Gene Aliases
FHX
Chromosome Location
Chr.12: 8032680 - 8055522 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018416.2 NP_060886.1 4 - 5 1568 79
XM_011520760.2 XP_011519062.1 4 - 5 1651 79
XM_011520761.2 XP_011519063.1 5 - 6 1752 79
XM_017019640.1 XP_016875129.1 3 - 4 315 79
GenBank mRNA AF155132.1 - 4 - 5 972 79
AF155133.1 - 4 - 5 972 79
BC126396.1 - 4 - 5 538 79
BC136305.1 - 4 - 5 538 79

View Full Product Details