The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018416.2 | NP_060886.1 | 5 - 6 | 1705 | 64 |
XM_011520760.2 | XP_011519062.1 | 5 - 6 | 1788 | 64 | |
XM_011520761.2 | XP_011519063.1 | 6 - 7 | 1889 | 64 | |
XM_017019640.1 | XP_016875129.1 | 4 - 5 | 452 | 64 | |
GenBank mRNA | AF155132.1 | - | 5 - 6 | 1109 | 64 |
AF155133.1 | - | 5 - 6 | 1109 | 64 | |
BC126396.1 | - | 5 - 6 | 675 | 64 | |
BC136305.1 | - | 5 - 6 | 675 | 64 |