The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018416.2 | NP_060886.1 | 7 - 8 | 2308 | 101 |
XM_011520760.2 | XP_011519062.1 | 7 - 8 | 2391 | 101 | |
XM_011520761.2 | XP_011519063.1 | 8 - 9 | 2492 | 101 | |
XM_017019640.1 | XP_016875129.1 | 6 - 7 | 1055 | 101 | |
GenBank mRNA | AF155132.1 | - | 7 - 8 | 1712 | 101 |
AF155133.1 | - | 7 - 8 | 1712 | 101 | |
BC126396.1 | - | 7 - 8 | 1278 | 101 | |
BC136305.1 | - | 7 - 8 | 1278 | 101 |