The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242307.1 | NP_001229236.1 | 11 - 12 | 1315 | 118 |
NM_001242309.1 | NP_001229238.1 | 10 - 11 | 1212 | 118 | |
NM_014889.3 | NP_055704.2 | 11 - 12 | 1315 | 118 | |
XM_005252345.3 | XP_005252402.1 | 11 - 12 | 1315 | 118 | |
XM_017015470.1 | XP_016870959.1 | 11 - 12 | 1383 | 118 | |
XM_017015471.1 | XP_016870960.1 | 11 - 12 | 1383 | 118 | |
XM_017015472.1 | XP_016870961.1 | 11 - 12 | 1212 | 118 | |
XM_017015473.1 | XP_016870962.1 | 11 - 12 | 1212 | 118 | |
GenBank mRNA | AB029027.2 | - | 1 - 2 | 44 | 118 |
AK002061.1 | - | 11 - 12 | 1283 | 118 | |
AK023476.1 | - | 4 - 5 | 430 | 118 | |
AK225084.1 | - | 11 - 12 | 1253 | 118 | |
AK294865.1 | - | 10 - 11 | 1228 | 118 | |
AK299464.1 | - | 10 - 11 | 1061 | 118 | |
AK303406.1 | - | 10 - 11 | 1153 | 118 | |
AK315117.1 | - | 11 - 12 | 1253 | 118 | |
BC001150.1 | - | 5 - 6 | 615 | 118 | |
BC005025.1 | - | 11 - 12 | 1250 | 118 | |
BC045577.1 | - | 11 - 12 | 1725 | 118 | |
BC095422.1 | - | 11 - 12 | 1256 | 118 | |
BC111987.1 | - | 11 - 12 | 1271 | 118 | |
BC113369.1 | - | 11 - 12 | 1271 | 118 | |
BM737694.1 | - | 5 - 6 | 543 | 118 | |
BX649017.1 | - | 11 - 12 | 1257 | 118 | |
CR749279.1 | - | 9 - 10 | 991 | 118 |