The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001015880.1 | NP_001015880.1 | 10 - 11 | 1487 | 69 |
NM_004670.3 | NP_004661.2 | 9 - 10 | 1472 | 69 | |
GenBank mRNA | AF074331.1 | - | 9 - 10 | 1272 | 69 |
AF091242.1 | - | 9 - 10 | 1289 | 69 | |
AF173365.1 | - | 10 - 11 | 1272 | 69 | |
AF313907.1 | - | 9 - 10 | 1209 | 69 | |
AK301453.1 | - | 9 - 10 | 1259 | 69 | |
AK312556.1 | - | 9 - 10 | 1313 | 69 | |
BC009894.2 | - | 9 - 10 | 1320 | 69 |