The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242413.2 | NP_001229342.1 | 10 - 11 | 1117 | 95 |
NM_001282644.1 | NP_001269573.1 | 10 - 11 | 1115 | 95 | |
NM_001282645.1 | NP_001269574.1 | 9 - 10 | 1056 | 95 | |
NM_001323265.1 | NP_001310194.1 | 10 - 11 | 1172 | 95 | |
NM_001323266.1 | NP_001310195.1 | 9 - 10 | 1054 | 95 | |
NM_001323267.1 | NP_001310196.1 | 10 - 11 | 1115 | 95 | |
NM_006257.4 | NP_006248.1 | 10 - 11 | 1117 | 95 | |
XM_005252496.4 | XP_005252553.1 | 10 - 11 | 1217 | 95 | |
XM_005252497.4 | XP_005252554.1 | 10 - 11 | 1219 | 95 | |
XM_006717465.3 | XP_006717528.1 | 12 - 13 | 3612 | 95 | |
XM_017016410.1 | XP_016871899.1 | 11 - 12 | 3523 | 95 | |
XM_017016411.1 | XP_016871900.1 | 11 - 12 | 3551 | 95 | |
GenBank mRNA | AK293935.1 | - | 9 - 10 | 994 | 95 |
AK308099.1 | - | 10 - 11 | 1090 | 95 | |
AK312411.1 | - | 10 - 11 | 1067 | 95 | |
AY702977.1 | - | 9 - 10 | 1017 | 95 | |
BC101465.1 | - | 9 - 10 | 1036 | 95 | |
BC113359.1 | - | 9 - 10 | 1036 | 95 | |
BX647657.1 | - | 9 - 10 | 1202 | 95 | |
L01087.1 | - | 10 - 11 | 1111 | 95 | |
L07032.1 | - | 9 - 10 | 1038 | 95 |