Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C19orf71
Entrez Gene ID
100128569
Gene Name
chromosome 19 open reading frame 71
Gene Aliases
-
Chromosome Location
Chr.19: 3539157 - 3544030 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135580.1 NP_001129052.1 3 - 4 468 87
XM_011527596.2 XP_011525898.1 3 - 4 394 87
GenBank mRNA AI138384.1 - 1 - 2 87
BX396051.2 - 3 - 4 472 86
DY654838.1 - 1 - 2 87

View Full Product Details