The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017973.1 | NP_001017973.1 | 14 - 15 | 2093 | 68 |
NM_001017974.1 | NP_001017974.1 | 14 - 15 | 1621 | 68 | |
NM_001142598.1 | NP_001136070.1 | 15 - 16 | 1757 | 68 | |
NM_001142599.1 | NP_001136071.1 | 15 - 16 | 1763 | 68 | |
NM_004199.2 | NP_004190.1 | 14 - 15 | 2099 | 68 | |
XM_005272116.4 | XP_005272173.1 | 14 - 15 | 1627 | 68 | |
XM_005272117.4 | XP_005272174.1 | 15 - 16 | 1785 | 68 | |
XM_005272118.4 | XP_005272175.1 | 15 - 16 | 1914 | 68 | |
XM_005272119.4 | XP_005272176.1 | 15 - 16 | 2104 | 68 | |
XM_005272120.4 | XP_005272177.1 | 15 - 16 | 1884 | 68 | |
XM_006714728.3 | XP_006714791.1 | 15 - 16 | 2161 | 68 | |
XM_006714729.3 | XP_006714792.1 | 15 - 16 | 1779 | 68 | |
XM_006714730.3 | XP_006714793.1 | 15 - 16 | 2155 | 68 | |
XM_011543705.2 | XP_011542007.1 | 12 - 13 | 1376 | 68 | |
XM_017010011.1 | XP_016865500.1 | 15 - 16 | 1878 | 68 | |
XM_017010012.1 | XP_016865501.1 | 13 - 14 | 1444 | 68 | |
GenBank mRNA | AY358970.1 | - | 14 - 15 | 1695 | 68 |
BC017062.1 | - | 14 - 15 | 1722 | 68 | |
BC035813.1 | - | 14 - 15 | 1590 | 68 | |
BX647924.1 | - | 14 - 15 | 1613 | 68 | |
U90441.1 | - | 14 - 15 | 1722 | 68 |