The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017973.1 | NP_001017973.1 | 5 - 6 | 1034 | 64 |
NM_001017974.1 | NP_001017974.1 | 5 - 6 | 562 | 64 | |
NM_001142598.1 | NP_001136070.1 | 6 - 7 | 698 | 64 | |
NM_001142599.1 | NP_001136071.1 | 6 - 7 | 698 | 64 | |
NM_004199.2 | NP_004190.1 | 5 - 6 | 1034 | 64 | |
XM_005272116.4 | XP_005272173.1 | 5 - 6 | 562 | 64 | |
XM_005272117.4 | XP_005272174.1 | 6 - 7 | 720 | 64 | |
XM_005272118.4 | XP_005272175.1 | 6 - 7 | 849 | 64 | |
XM_005272119.4 | XP_005272176.1 | 6 - 7 | 1039 | 64 | |
XM_005272120.4 | XP_005272177.1 | 6 - 7 | 819 | 64 | |
XM_006714728.3 | XP_006714791.1 | 6 - 7 | 1096 | 64 | |
XM_006714729.3 | XP_006714792.1 | 6 - 7 | 720 | 64 | |
XM_006714730.3 | XP_006714793.1 | 6 - 7 | 1096 | 64 | |
XM_011543705.2 | XP_011542007.1 | 4 - 5 | 488 | 64 | |
XM_017010011.1 | XP_016865500.1 | 6 - 7 | 819 | 64 | |
XM_017010012.1 | XP_016865501.1 | 5 - 6 | 562 | 64 | |
GenBank mRNA | AU117989.1 | - | 5 - 6 | 562 | 64 |
AY358970.1 | - | 5 - 6 | 636 | 64 | |
BC017062.1 | - | 5 - 6 | 663 | 64 | |
BC035813.1 | - | 5 - 6 | 531 | 64 | |
BI758477.1 | - | 6 - 7 | 649 | 64 | |
BX647924.1 | - | 5 - 6 | 554 | 64 | |
U90441.1 | - | 5 - 6 | 657 | 64 |